Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAGTTAGCTAATGGCATCCCTTC[C/T]ATCTTTTCATCACTGTGGCTTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611736 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GREB1 - growth regulation by estrogen in breast cancer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014668.3 | Intron | NP_055483.2 | ||||
NM_033090.2 | Intron | NP_149081.1 | ||||
NM_148903.2 | Intron | NP_683701.2 | ||||
XM_005246192.4 | Intron | XP_005246249.1 | ||||
XM_005246196.3 | Intron | XP_005246253.1 | ||||
XM_011510418.2 | Intron | XP_011508720.1 | ||||
XM_011510419.2 | Intron | XP_011508721.1 | ||||
XM_011510422.2 | Intron | XP_011508724.1 | ||||
XM_011510423.2 | Intron | XP_011508725.1 |
MIR4429 - microRNA 4429 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |