Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGAGAGATTAGTTTCAGAAAGTT[C/T]CATCTAAGATGTTTGTGATTAGAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604002 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROCK2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ROCK2 - Rho associated coiled-coil containing protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321643.1 | Intron | NP_001308572.1 | ||||
NM_004850.4 | Intron | NP_004841.2 | ||||
XM_005246190.3 | Intron | XP_005246247.1 | ||||
XM_011510417.2 | Intron | XP_011508719.1 | ||||
XM_017005378.1 | Intron | XP_016860867.1 | ||||
XM_017005379.1 | Intron | XP_016860868.1 |
Set Membership: |
HapMap |