Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACATTCACTGTACATTAATTTC[A/G]ACAGTAATACCGGCACCAGTTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604002 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PQLC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PQLC3 - PQ loop repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282710.1 | 7114 | Intron | NP_001269639.1 | |||
NM_001282711.1 | 7114 | Intron | NP_001269640.1 | |||
NM_001282712.1 | 7114 | Intron | NP_001269641.1 | |||
NM_152391.4 | 7114 | Intron | NP_689604.1 |
ROCK2 - Rho associated coiled-coil containing protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321643.1 | 7114 | UTR 3 | NP_001308572.1 | |||
NM_004850.4 | 7114 | UTR 3 | NP_004841.2 | |||
XM_005246190.3 | 7114 | UTR 3 | XP_005246247.1 | |||
XM_011510417.2 | 7114 | UTR 3 | XP_011508719.1 | |||
XM_017005378.1 | 7114 | Intron | XP_016860867.1 | |||
XM_017005379.1 | 7114 | UTR 3 | XP_016860868.1 |
Set Membership: |
HapMap |