Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAAAGGACTAACAATGAACTGA[A/G]AAGGTGACCAAAAATCATTCCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176887 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTPN2 - protein tyrosine phosphatase, non-receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207013.1 | Intron | NP_001193942.1 | ||||
NM_001308287.1 | Intron | NP_001295216.1 | ||||
NM_002828.3 | Intron | NP_002819.2 | ||||
NM_080422.2 | Intron | NP_536347.1 | ||||
NM_080423.2 | Intron | NP_536348.1 | ||||
XM_005258124.3 | Intron | XP_005258181.1 | ||||
XM_005258125.3 | Intron | XP_005258182.1 | ||||
XM_011525705.2 | Intron | XP_011524007.1 | ||||
XM_011525706.2 | Intron | XP_011524008.1 | ||||
XM_011525707.2 | Intron | XP_011524009.1 | ||||
XM_017025884.1 | Intron | XP_016881373.1 | ||||
XM_017025885.1 | Intron | XP_016881374.1 | ||||
XM_017025886.1 | Intron | XP_016881375.1 | ||||
XM_017025887.1 | Intron | XP_016881376.1 | ||||
XM_017025888.1 | Intron | XP_016881377.1 |
Set Membership: |
HapMap |