Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCATCGGAGCAGCTGTGGGAGCC[A/G]CAGATGATGCAGGCTATGTCTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609749 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UXS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
UXS1 - UDP-glucuronate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253875.1 | Intron | NP_001240804.1 | ||||
NM_001253876.1 | Intron | NP_001240805.1 | ||||
NM_025076.4 | Intron | NP_079352.2 | ||||
XM_006712775.3 | Intron | XP_006712838.1 | ||||
XM_011511902.2 | Intron | XP_011510204.2 | ||||
XM_011511903.2 | Intron | XP_011510205.2 | ||||
XM_011511904.2 | Intron | XP_011510206.1 | ||||
XM_017005008.1 | Intron | XP_016860497.1 | ||||
XM_017005009.1 | Intron | XP_016860498.1 | ||||
XM_017005010.1 | Intron | XP_016860499.1 | ||||
XM_017005011.1 | Intron | XP_016860500.1 | ||||
XM_017005012.1 | Intron | XP_016860501.1 | ||||
XM_017005013.1 | Intron | XP_016860502.1 | ||||
XM_017005014.1 | Intron | XP_016860503.1 |
Set Membership: |
HapMap |