Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAGAGGTGAGGCAGGATGCGGGT[C/G]AGGCCCACGCTGGGCACAGGCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602038 | ||||||||||||||||||||
Literature Links: |
DUSP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP8 - dual specificity phosphatase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004420.2 | 1638 | Silent Mutation | CTC,CTG | L,L 161 | NP_004411.2 | |
XM_011519932.2 | 1638 | Silent Mutation | CTC,CTG | L,L 161 | XP_011518234.1 | |
XM_011519933.2 | 1638 | Silent Mutation | CTC,CTG | L,L 161 | XP_011518235.1 |
LOC101927562 - uncharacterized LOC101927562 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |