Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAATAGTGAGCGTTGACTGACCA[C/T]TGCTTTCAAAGTGAGTTGTGTTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614572 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP42 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZFP42 - ZFP42 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304358.1 | Intron | NP_001291287.1 | ||||
NM_174900.4 | Intron | NP_777560.2 | ||||
XM_011531604.2 | Intron | XP_011529906.1 | ||||
XM_011531605.1 | Intron | XP_011529907.1 | ||||
XM_011531606.1 | Intron | XP_011529908.1 | ||||
XM_011531607.2 | Intron | XP_011529909.1 |
Set Membership: |
HapMap |