Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCTGCAATTTTCTTTGTGAACA[C/T]CTGCAAGACTCTGCATGGTCCAGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 138590 | ||||||||||||||||||||
Literature Links: |
GYPE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GYPE - glycophorin E (MNS blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002102.3 | Intron | NP_002093.2 | ||||
NM_198682.2 | Intron | NP_941391.2 | ||||
XM_017008139.1 | Intron | XP_016863628.1 | ||||
XM_017008140.1 | Intron | XP_016863629.1 |