Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGAGATGTATGCACACACAAGT[A/C]AACTAGAGAGAATTTTATTCTATGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605011 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADAMTS3 - ADAM metallopeptidase with thrombospondin type 1 motif 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014243.2 | 6147 | UTR 3 | NP_055058.2 | |||
XM_011532421.2 | 6147 | UTR 3 | XP_011530723.1 | |||
XM_011532422.2 | 6147 | UTR 3 | XP_011530724.1 |
Set Membership: |
HapMap |