Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAACATGAATTGTTAAAAGGTAA[C/T]GAAACTTTAGGCTAGGGTCAGTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605200 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HERC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HERC3 - HECT and RLD domain containing E3 ubiquitin protein ligase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271602.1 | Intron | NP_001258531.1 | ||||
NM_001318505.1 | Intron | NP_001305434.1 | ||||
NM_014606.2 | Intron | NP_055421.1 | ||||
XM_005263327.3 | Intron | XP_005263384.1 | ||||
XM_005263329.3 | Intron | XP_005263386.1 | ||||
XM_005263330.3 | Intron | XP_005263387.1 | ||||
XM_005263331.1 | Intron | XP_005263388.1 | ||||
XM_005263332.3 | Intron | XP_005263389.1 | ||||
XM_011532397.2 | Intron | XP_011530699.1 | ||||
XM_017008806.1 | Intron | XP_016864295.1 | ||||
XM_017008807.1 | Intron | XP_016864296.1 | ||||
XM_017008808.1 | Intron | XP_016864297.1 |
Set Membership: |
HapMap |