Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTGTCAGAAAGGAAAATATATC[C/T]CACTTCTCTCCTTGTAGACATCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR186 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR186 - microRNA 186 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZRANB2 - zinc finger RANBP2-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005455.4 | 1910 | UTR 3 | NP_005446.2 | |||
NM_203350.2 | 1910 | UTR 3 | NP_976225.1 |
ZRANB2-AS1 - ZRANB2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |