Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCGACTCACCATTTGGATGGTG[C/G]CTGAATCCACATTCAGTTCCTGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604468 MIM: 608042 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAP3K6 - mitogen-activated protein kinase kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297609.1 | 3943 | Missense Mutation | GCC,GGC | A,G 1225 | NP_001284538.1 | |
NM_004672.4 | 3943 | Missense Mutation | GCC,GGC | A,G 1233 | NP_004663.3 | |
XM_017002771.1 | 3943 | Missense Mutation | GCC,GGC | A,G 814 | XP_016858260.1 | |
XM_017002772.1 | 3943 | Intron | XP_016858261.1 |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |