Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGCATTACATAGTCTTTCAAAA[A/G]AGCTGGGCAGCTGGAGAAGTGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182360 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP12A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP12A - ATPase H+/K+ transporting non-gastric alpha2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185085.1 | Intron | NP_001172014.1 | ||||
NM_001676.5 | Intron | NP_001667.4 |
Set Membership: |
HapMap |