Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATTAAACACGGAAATGGATTTC[A/G]AGATGTGTGCATGTACACAAAAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114080 MIM: 607049 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMK4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CAMK4 - calcium/calmodulin dependent protein kinase IV | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323374.1 | 1645 | Intron | NP_001310303.1 | |||
NM_001323375.1 | 1645 | Intron | NP_001310304.1 | |||
NM_001323376.1 | 1645 | Intron | NP_001310305.1 | |||
NM_001323377.1 | 1645 | Intron | NP_001310306.1 | |||
NM_001744.5 | 1645 | Intron | NP_001735.1 |
STARD4 - StAR related lipid transfer domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308056.1 | 1645 | UTR 3 | NP_001294985.1 | |||
NM_001308057.1 | 1645 | Intron | NP_001294986.1 | |||
NM_001308058.1 | 1645 | Intron | NP_001294987.1 | |||
NM_001308059.1 | 1645 | UTR 3 | NP_001294988.1 | |||
NM_001308060.1 | 1645 | UTR 3 | NP_001294989.1 | |||
NM_001308061.1 | 1645 | UTR 3 | NP_001294990.1 | |||
NM_139164.2 | 1645 | UTR 3 | NP_631903.1 | |||
XM_017009041.1 | 1645 | UTR 3 | XP_016864530.1 | |||
XM_017009042.1 | 1645 | UTR 3 | XP_016864531.1 | |||
XM_017009043.1 | 1645 | UTR 3 | XP_016864532.1 | |||
XM_017009044.1 | 1645 | UTR 3 | XP_016864533.1 |
Set Membership: |
HapMap |