Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATAAAATCACCTGAAATGGAA[C/T]GGAATGGCTACAGCGAGTGTCCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138280 MIM: 600555 | ||||||||||||||||||||||||||||||||
Literature Links: |
GLS PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GLS - glutaminase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT1 - signal transducer and activator of transcription 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007315.3 | Intron | NP_009330.1 | ||||
NM_139266.2 | Intron | NP_644671.1 | ||||
XM_006712718.1 | Intron | XP_006712781.1 | ||||
XM_017004783.1 | Intron | XP_016860272.1 |
Set Membership: |
HapMap |