Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAGATTCACTTCTTACACAAT[C/T]CATTGTTGCTATAATTCAAACCCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614738 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MPC1 - mitochondrial pyruvate carrier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270879.1 | Intron | NP_001257808.1 | ||||
NM_016098.3 | Intron | NP_057182.1 | ||||
XM_011535894.1 | Intron | XP_011534196.1 | ||||
XM_011535895.1 | Intron | XP_011534197.1 |