Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCTGGAGGCGAAGTTCAGGTC[A/G]TTATTGTTCTCTGTGTATAGAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615358 MIM: 609390 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK9 - adenylate kinase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FIG4 - FIG4 phosphoinositide 5-phosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014845.5 | 190 | Intron | NP_055660.1 | |||
XM_011536281.2 | 190 | UTR 5 | XP_011534583.1 | |||
XM_017011591.1 | 190 | Intron | XP_016867080.1 | |||
XM_017011592.1 | 190 | Intron | XP_016867081.1 | |||
XM_017011593.1 | 190 | Intron | XP_016867082.1 |
Set Membership: |
HapMap |