Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAATGCCTTGTCCAGTGGCACTT[A/G]ATACTAAACTCCATCAGATTCTCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610564 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDSS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDSS2 - prenyl (decaprenyl) diphosphate synthase, subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020381.3 | Intron | NP_065114.3 | ||||
XM_011535956.2 | Intron | XP_011534258.1 | ||||
XM_011535957.2 | Intron | XP_011534259.1 | ||||
XM_011535958.2 | Intron | XP_011534260.1 | ||||
XM_011535959.2 | Intron | XP_011534261.1 | ||||
XM_011535960.2 | Intron | XP_011534262.1 | ||||
XM_011535961.2 | Intron | XP_011534263.1 | ||||
XM_011535962.2 | Intron | XP_011534264.1 | ||||
XM_011535963.2 | Intron | XP_011534265.1 | ||||
XM_017011082.1 | Intron | XP_016866571.1 |