Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACAATGGCGGTCGCTCGTGCAGC[C/T]TTGGGGCCATTGGTGACGGGTCTGT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604605 MIM: 613891 | |||||||||||||||||||||||
Literature Links: |
KALRN PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KALRN - kalirin, RhoGEF kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR544B - microRNA 544b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UMPS - uridine monophosphate synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000373.3 | 127 | Silent Mutation | GCC,GCT | A,A 7 | NP_000364.1 |