Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATAGACTTTAGCACCCACTTT[C/G]TGTTCACCCTGCCAACACTGTCTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614719 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCMF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
KCMF1 - potassium channel modulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020122.4 | Intron | NP_064507.3 | ||||
XM_006712052.3 | Intron | XP_006712115.1 | ||||
XM_011532990.1 | Intron | XP_011531292.1 | ||||
XM_017004511.1 | Intron | XP_016860000.1 |
Set Membership: |
HapMap |