Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGGGGTTTGGATGTTCTCAAG[G/T]AGGCAACAGAGTTTTTTTGTTTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC142 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC142 - coiled-coil domain containing 142 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC31 - tetratricopeptide repeat domain 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022492.4 | Intron | NP_071937.4 | ||||
XM_005264492.1 | Intron | XP_005264549.1 | ||||
XM_011533035.1 | Intron | XP_011531337.1 | ||||
XM_011533036.1 | Intron | XP_011531338.1 | ||||
XM_011533037.1 | Intron | XP_011531339.1 | ||||
XM_011533040.1 | Intron | XP_011531342.1 | ||||
XM_017004706.1 | Intron | XP_016860195.1 | ||||
XM_017004707.1 | Intron | XP_016860196.1 | ||||
XM_017004708.1 | Intron | XP_016860197.1 | ||||
XM_017004709.1 | Intron | XP_016860198.1 |
Set Membership: |
HapMap |