Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGGGGCGCGTCGTCGCGGAGCT[C/T]GTCTCCTCGCTGCTGGGGTTGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610995 MIM: 603518 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1285-2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR1285-2 - microRNA 1285-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCYOX1 - prenylcysteine oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016297.3 | 90 | Silent Mutation | CTC,CTT | L,L 8 | NP_057381.3 |
TIA1 - TIA1 cytotoxic granule-associated RNA binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |