Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTACAGAATACTGACCATGTTT[A/G]CCTGAAGAGATAGGAACCACCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607788 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCFD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCFD2 - multiple coagulation factor deficiency 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171506.2 | 3237 | Intron | NP_001164977.1 | |||
NM_001171507.2 | 3237 | UTR 3 | NP_001164978.1 | |||
NM_001171508.2 | 3237 | UTR 3 | NP_001164979.1 | |||
NM_001171509.2 | 3237 | Intron | NP_001164980.1 | |||
NM_001171510.2 | 3237 | Intron | NP_001164981.1 | |||
NM_001171511.2 | 3237 | UTR 3 | NP_001164982.1 | |||
NM_139279.5 | 3237 | Intron | NP_644808.1 |
Set Membership: |
HapMap |