Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTGCAGGTTTCTAGAGGCCTTC[A/G]GAGAGCATCAGCTATTTGTGATGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609359 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS1BP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HS1BP3 - HCLS1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022460.3 | 1624 | Intron | NP_071905.3 | |||
XM_017004696.1 | 1624 | Intron | XP_016860185.1 | |||
XM_017004697.1 | 1624 | UTR 3 | XP_016860186.1 | |||
XM_017004698.1 | 1624 | Intron | XP_016860187.1 | |||
XM_017004699.1 | 1624 | Intron | XP_016860188.1 | |||
XM_017004700.1 | 1624 | Intron | XP_016860189.1 | |||
XM_017004701.1 | 1624 | Intron | XP_016860190.1 |