Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTAACTTCATACATTTATCGGCA[C/T]TGTCCAAAATATTTTATTCTTTAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611099 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1C2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP6V1C2 - ATPase H+ transporting V1 subunit C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039362.1 | 1857 | UTR 3 | NP_001034451.1 | |||
NM_144583.3 | 1857 | UTR 3 | NP_653184.2 | |||
XM_011510339.2 | 1857 | UTR 3 | XP_011508641.1 | |||
XM_011510340.2 | 1857 | UTR 3 | XP_011508642.1 | |||
XM_011510341.2 | 1857 | Intron | XP_011508643.1 | |||
XM_017003745.1 | 1857 | UTR 3 | XP_016859234.1 |
PDIA6 - protein disulfide isomerase family A member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282704.1 | 1857 | UTR 3 | NP_001269633.1 | |||
NM_001282705.1 | 1857 | UTR 3 | NP_001269634.1 | |||
NM_001282706.1 | 1857 | UTR 3 | NP_001269635.1 | |||
NM_001282707.1 | 1857 | UTR 3 | NP_001269636.1 | |||
NM_005742.3 | 1857 | UTR 3 | NP_005733.1 | |||
XM_011510308.1 | 1857 | UTR 3 | XP_011508610.1 |
Set Membership: |
HapMap |