Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179530 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAP1B - RAP1B, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010942.2 | Intron | NP_001010942.1 | ||||
NM_001251917.1 | Intron | NP_001238846.1 | ||||
NM_001251918.1 | Intron | NP_001238847.1 | ||||
NM_001251921.1 | Intron | NP_001238850.1 | ||||
NM_001251922.1 | Intron | NP_001238851.1 | ||||
NM_015646.5 | Intron | NP_056461.1 |
SNORA70G - small nucleolar RNA, H/ACA box 70G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |