Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGGGACAGATTATATCATTCC[G/T]GAAGGCATATACCTTTTGTTGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176882 MIM: 602853 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTPRB - protein tyrosine phosphatase, receptor type B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719528.1 | 2880 | Intron | XP_006719591.1 | |||
XM_006719529.3 | 2880 | Intron | XP_006719592.1 | |||
XM_011538614.1 | 2880 | Intron | XP_011536916.1 | |||
XM_017019723.1 | 2880 | Intron | XP_016875212.1 | |||
XM_017019724.1 | 2880 | Intron | XP_016875213.1 |
PTPRR - protein tyrosine phosphatase, receptor type R | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207015.1 | 2880 | UTR 3 | NP_001193944.1 | |||
NM_001207016.1 | 2880 | UTR 3 | NP_001193945.1 | |||
NM_002849.3 | 2880 | UTR 3 | NP_002840.2 | |||
NM_130846.2 | 2880 | UTR 3 | NP_570897.2 | |||
XM_011538615.2 | 2880 | UTR 3 | XP_011536917.1 |
Set Membership: |
HapMap |