Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATGCCTATGGAATGCTTTGGCC[C/T]GGTATACAGCAAATGCTCATTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609429 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXN4 - forkhead box N4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213596.2 | Intron | NP_998761.2 | ||||
XM_011537922.2 | Intron | XP_011536224.1 | ||||
XM_011537923.2 | Intron | XP_011536225.1 | ||||
XM_017018818.1 | Intron | XP_016874307.1 | ||||
XM_017018819.1 | Intron | XP_016874308.1 |