Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCTTAAAATAGGTAACTATAAT[A/G]GATTACTAAGAACAAAGTAACAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616013 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4orf17 - chromosome 4 open reading frame 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRMT10A - tRNA methyltransferase 10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134665.2 | 2565 | UTR 3 | NP_001128137.1 | |||
NM_001134666.2 | 2565 | UTR 3 | NP_001128138.1 | |||
NM_152292.4 | 2565 | UTR 3 | NP_689505.1 | |||
XM_005263352.3 | 2565 | UTR 3 | XP_005263409.1 | |||
XM_006714417.2 | 2565 | UTR 3 | XP_006714480.1 |