Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTGAAACCATTTTGCAGATGTC[A/G]GTCCTGAGGTACTAAGAGGTTTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605768 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAPP1 - dual adaptor of phosphotyrosine and 3-phosphoinositides 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306151.1 | Intron | NP_001293080.1 | ||||
NM_014395.2 | Intron | NP_055210.2 | ||||
XM_011531840.2 | Intron | XP_011530142.1 | ||||
XM_011531842.1 | Intron | XP_011530144.1 | ||||
XM_011531843.2 | Intron | XP_011530145.1 | ||||
XM_017008023.1 | Intron | XP_016863512.1 | ||||
XM_017008024.1 | Intron | XP_016863513.1 |