Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCCAATTCAAGTTTTTCAGTAC[C/T]CTAAATTCTTCAGGGCCAACAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNAJB14 - DnaJ heat shock protein family (Hsp40) member B14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031723.3 | Intron | NP_001026893.1 | ||||
NM_001278310.1 | Intron | NP_001265239.1 | ||||
NM_001278311.1 | Intron | NP_001265240.1 | ||||
XM_011532262.2 | Intron | XP_011530564.1 | ||||
XM_011532263.2 | Intron | XP_011530565.1 | ||||
XM_011532265.2 | Intron | XP_011530567.1 | ||||
XM_017008628.1 | Intron | XP_016864117.1 |