Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATACTTTTGGACTGATGTACAA[C/T]TCTCATCTCTATAATGGAAATAAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615912 MIM: 611355 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTCD PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSTCD - glutathione S-transferase C-terminal domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031720.3 | Intron | NP_001026890.2 | ||||
NM_024751.3 | Intron | NP_079027.2 | ||||
XM_005263222.3 | Intron | XP_005263279.1 | ||||
XM_011532248.2 | Intron | XP_011530550.1 | ||||
XM_011532249.2 | Intron | XP_011530551.1 | ||||
XM_011532252.2 | Intron | XP_011530554.1 |
INTS12 - integrator complex subunit 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |