Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCACAAACGCTTTGGCAATCAG[A/G]TAAAGACTATGAATATTGTGTCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616899 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBCK PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TBCK - TBC1 domain containing kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163435.2 | Intron | NP_001156907.1 | ||||
NM_001163436.2 | Intron | NP_001156908.1 | ||||
NM_001163437.2 | Intron | NP_001156909.1 | ||||
NM_001290768.1 | Intron | NP_001277697.1 | ||||
NM_033115.4 | Intron | NP_149106.2 | ||||
XM_011532417.2 | Intron | XP_011530719.1 | ||||
XM_017008846.1 | Intron | XP_016864335.1 | ||||
XM_017008847.1 | Intron | XP_016864336.1 | ||||
XM_017008848.1 | Intron | XP_016864337.1 | ||||
XM_017008849.1 | Intron | XP_016864338.1 |
Set Membership: |
HapMap |