Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACTCCAAAGTATGATTTTTATCT[C/G]CCTAACTCTATGAATCTAGCAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613975 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI44L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFI44L - interferon induced protein 44 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006820.3 | Intron | NP_006811.2 | ||||
XM_005270391.3 | Intron | XP_005270448.1 | ||||
XM_005270392.4 | Intron | XP_005270449.1 | ||||
XM_005270393.4 | Intron | XP_005270450.1 | ||||
XM_006710304.3 | Intron | XP_006710367.1 | ||||
XM_011540539.2 | Intron | XP_011538841.1 | ||||
XM_017000120.1 | Intron | XP_016855609.1 |