Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAATTTTCCCTTAGAGTTCACC[A/G]AACAGCAGAAAGAATTTCAAGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607008 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACADM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACADM - acyl-CoA dehydrogenase, C-4 to C-12 straight chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000016.5 | 480 | Missense Mutation | AAA,GAA | K,E 43 | NP_000007.1 | |
NM_001127328.2 | 480 | Missense Mutation | AAA,GAA | K,E 47 | NP_001120800.1 | |
NM_001286042.1 | 480 | Missense Mutation | AAA,GAA | K,E 7 | NP_001272971.1 | |
NM_001286043.1 | 480 | Missense Mutation | AAA,GAA | K,E 43 | NP_001272972.1 | |
NM_001286044.1 | 480 | UTR 5 | NP_001272973.1 |
DLSTP1 - dihydrolipoamide S-succinyltransferase pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC44A5 - solute carrier family 44 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |