Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCCAGGAGCCATCTAAAGGTA[A/C]AAACTGAGAAAGATTTAACTGAATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610804 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35D1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC35D1 - solute carrier family 35 member D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015139.2 | 5960 | UTR 3 | NP_055954.1 | |||
XM_006710478.1 | 5960 | UTR 3 | XP_006710541.1 | |||
XM_011541070.1 | 5960 | UTR 3 | XP_011539372.1 |
Set Membership: |
HapMap |