Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGTTTACTCCAGCTGTCTTATGA[C/T]TAGGAAAAAGGAAACATAAAAATTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610785 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDIK1L PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDIK1L - PDLIM1 interacting kinase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243532.1 | Intron | NP_001230461.1 | ||||
NM_001243533.1 | Intron | NP_001230462.1 | ||||
NM_152835.4 | Intron | NP_690048.1 | ||||
XM_005245743.4 | Intron | XP_005245800.1 | ||||
XM_006710381.3 | Intron | XP_006710444.1 | ||||
XM_011540792.2 | Intron | XP_011539094.1 |
Set Membership: |
HapMap |