Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCAGAGTTAGAAACAGCACCTA[A/G]CCTATTTTTATATCCAGCGAATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610323 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MTDH - metadherin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178812.3 | Intron | NP_848927.2 | ||||
XM_005251099.3 | Intron | XP_005251156.1 | ||||
XM_011517367.2 | Intron | XP_011515669.1 | ||||
XM_011517368.2 | Intron | XP_011515670.1 | ||||
XM_011517369.2 | Intron | XP_011515671.1 | ||||
XM_011517370.2 | Intron | XP_011515672.1 | ||||
XM_017013966.1 | Intron | XP_016869455.1 | ||||
XM_017013967.1 | Intron | XP_016869456.1 | ||||
XM_017013968.1 | Intron | XP_016869457.1 |