Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATATTAGACTGTCAGAGCTTAG[A/G]AAGTCCCCGATTGTCCATAGACGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIPAL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NIPAL2 - NIPA like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321635.1 | 1568 | UTR 3 | NP_001308564.1 | |||
NM_001321636.1 | 1568 | UTR 3 | NP_001308565.1 | |||
NM_024759.2 | 1568 | UTR 3 | NP_079035.1 | |||
XM_011517302.2 | 1568 | Intron | XP_011515604.1 |