Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTATTGAGGGCAGGGGCTGAAG[C/T]GGTGTTGTATCATAGCTTGTCTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606604 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO32 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXO32 - F-box protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242463.1 | 2657 | UTR 3 | NP_001229392.1 | |||
NM_058229.3 | 2657 | UTR 3 | NP_478136.1 | |||
NM_148177.2 | 2657 | UTR 3 | NP_680482.1 |