Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGGTCTACTGCCGACATAATAC[A/C]GTTTCATAAACATGAAAACATCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603745 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLIT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLIT3 - slit guidance ligand 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271946.1 | 5407 | UTR 3 | NP_001258875.1 | |||
NM_003062.3 | 5407 | UTR 3 | NP_003053.1 | |||
XM_017009779.1 | 5407 | UTR 3 | XP_016865268.1 |