Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATTAGTATAGTTGGTATAATA[A/C]CCATGGAATAAATAACCCTTTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC150 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCDC150 - coiled-coil domain containing 150 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080539.1 | Intron | NP_001074008.1 | ||||
XM_006712438.3 | Intron | XP_006712501.1 | ||||
XM_006712439.3 | Intron | XP_006712502.1 | ||||
XM_011510985.2 | Intron | XP_011509287.1 | ||||
XM_011510987.2 | Intron | XP_011509289.1 | ||||
XM_011510988.2 | Intron | XP_011509290.1 | ||||
XM_017003861.1 | Intron | XP_016859350.1 | ||||
XM_017003862.1 | Intron | XP_016859351.1 | ||||
XM_017003863.1 | Intron | XP_016859352.1 | ||||
XM_017003864.1 | Intron | XP_016859353.1 | ||||
XM_017003865.1 | Intron | XP_016859354.1 | ||||
XM_017003866.1 | Intron | XP_016859355.1 |