Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGGAAGGCTGGCTTAGGAATTA[C/T]CTTCCTGAGACTCTTCCTACTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100730 MIM: 605895 MIM: 612972 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNG - cholinergic receptor nicotinic gamma subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF4E2 - eukaryotic translation initiation factor 4E family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276336.1 | Intron | NP_001263265.1 | ||||
NM_001276337.1 | Intron | NP_001263266.1 | ||||
NM_001282958.1 | Intron | NP_001269887.1 | ||||
NM_004846.3 | Intron | NP_004837.1 | ||||
XM_005246975.3 | Intron | XP_005247032.1 | ||||
XM_011512206.2 | Intron | XP_011510508.1 | ||||
XM_017005377.1 | Intron | XP_016860866.1 |
MIR5001 - microRNA 5001 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIGD1 - tigger transposable element derived 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145702.2 | Intron | NP_663748.1 |