Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATGCAGCTGCTTTCAAATCATC[C/T]GAAAGACTTCTACATTCCTATACCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600814 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRE11A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRE11A - MRE11 homolog A, double strand break repair nuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005590.3 | 4098 | UTR 3 | NP_005581.2 | |||
NM_005591.3 | 4098 | UTR 3 | NP_005582.1 | |||
XM_005274008.3 | 4098 | UTR 3 | XP_005274065.1 | |||
XM_006718842.3 | 4098 | UTR 3 | XP_006718905.1 | |||
XM_011542837.2 | 4098 | UTR 3 | XP_011541139.1 | |||
XM_017017772.1 | 4098 | UTR 3 | XP_016873261.1 | |||
XM_017017773.1 | 4098 | Intron | XP_016873262.1 |