Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTACACAGTGCATTGTTTCTA[C/T]AGGATAGCTGTGATCAAACTGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601568 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADD3 - adducin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001121.3 | Intron | NP_001112.2 | ||||
NM_001320591.1 | Intron | NP_001307520.1 | ||||
NM_001320592.1 | Intron | NP_001307521.1 | ||||
NM_001320593.1 | Intron | NP_001307522.1 | ||||
NM_001320594.1 | Intron | NP_001307523.1 | ||||
NM_016824.4 | Intron | NP_058432.1 | ||||
NM_019903.4 | Intron | NP_063968.1 |
ADD3-AS1 - ADD3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |