Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAACTATTTACATGACTAGGCC[A/T]TTAGTTACCACTGTGGGATGATGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP58 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CFAP58 - cilia and flagella associated protein 58 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008723.1 | 434 | Intron | NP_001008723.1 | |||
XM_011539377.2 | 434 | Intron | XP_011537679.1 | |||
XM_017015787.1 | 434 | Intron | XP_016871276.1 | |||
XM_017015788.1 | 434 | UTR 5 | XP_016871277.1 | |||
XM_017015789.1 | 434 | Intron | XP_016871278.1 | |||
XM_017015790.1 | 434 | Intron | XP_016871279.1 |
CFAP58-AS1 - CFAP58 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |