Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAAGTGAATAATCAAGTCACAA[C/T]GGCTTACTTTGCATCACTAAGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RRP12 - ribosomal RNA processing 12 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145114.1 | Intron | NP_001138586.1 | ||||
NM_001284337.1 | Intron | NP_001271266.1 | ||||
NM_015179.3 | Intron | NP_055994.2 | ||||
XM_011539555.1 | Intron | XP_011537857.1 | ||||
XM_011539556.2 | Intron | XP_011537858.1 |