Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCCTCTGTTGGAAAAACAGAA[C/T]TGTCTGACTTATTGTGGCTGTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1287 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR1287 - microRNA 1287 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYROXD2 - pyridine nucleotide-disulphide oxidoreductase domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032709.2 | Intron | NP_116098.2 | ||||
XM_011540293.2 | Intron | XP_011538595.1 | ||||
XM_011540301.2 | Intron | XP_011538603.1 | ||||
XM_017016835.1 | Intron | XP_016872324.1 | ||||
XM_017016836.1 | Intron | XP_016872325.1 | ||||
XM_017016837.1 | Intron | XP_016872326.1 | ||||
XM_017016838.1 | Intron | XP_016872327.1 | ||||
XM_017016839.1 | Intron | XP_016872328.1 | ||||
XM_017016840.1 | Intron | XP_016872329.1 | ||||
XM_017016841.1 | Intron | XP_016872330.1 |
Set Membership: |
HapMap |