Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATAGGCTGATTATAGACCCTTC[C/T]AGTTTCAATTATCAATGTGAGACTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603594 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFSF4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TNFSF4 - tumor necrosis factor superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297562.1 | 2292 | UTR 3 | NP_001284491.1 | |||
NM_003326.4 | 2292 | UTR 3 | NP_003317.1 | |||
XM_011509964.2 | 2292 | UTR 3 | XP_011508266.2 | |||
XM_017002228.1 | 2292 | UTR 3 | XP_016857717.1 | |||
XM_017002229.1 | 2292 | UTR 3 | XP_016857718.1 | |||
XM_017002230.1 | 2292 | UTR 3 | XP_016857719.1 |