Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGATGTGCTACATGTTCATTA[C/T]GGTGCTAGCAAAAGTAGGTAGTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600522 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G4A - phospholipase A2 group IVA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311193.1 | Intron | NP_001298122.1 | ||||
NM_024420.2 | Intron | NP_077734.1 | ||||
XM_005245267.3 | Intron | XP_005245324.2 | ||||
XM_011509642.2 | Intron | XP_011507944.1 |